Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse Recombinant Vaspin (from HEK293 cells)
  102515-020
 :  Adipogen
Small Business Enterprise
 :  
Mouse Recombinant Vaspin (from HEK293 cells)
  102515-020
 :  Adipogen
 :  AG-40A-0094-C010
Small Business Enterprise
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Size:
    10 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1EU/µg
  • Gene ID:
    Q7TMF5
  • Protein Synonyms:
    OL-64|Serpin A12|Visceral Adipose Tissue-derived Serine Protease Inhibitor
  • Protein/Peptide Name:
    Vaspin
  • Purity:
    90%
  • Molecular Weight:
    ~50kDa (SDS-PAGE)
  • Sequence:
    21-413
  • Endotoxin Level:
    Low
  • Concentration:
    0.5mg/ml
  • Formulation:
    Liquid. 0.2µm-filtered solution in PBS, pH 7.2.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102515-020
  • Supplier no.:
    AG40A0094C010

 

 

Vaspin (Visceral adipose tissue-derived serpin; Serpin A12), a serine protease inhibitor (serpin), is an insulin-sensitizing adipocytokine that has been isolated from both visceral and subcutaneous white adipose tissue. Based on recent findings, vaspin is suggested to regulate immune responses and inflammation and was found to be correlated with various metabolic parameters. Vaspin may represent a novel biomarker for obesity and impaired insulin sensitivity and might serve as a new therapeutic target of metabolic syndrome.