About this item
Nucleobindin-2 (NUCB2; Prepronesfatin) was first isolated in adipocytes, but is also expressed by gastric mucosa and pancreatic beta cells. This pattern of nucleobindin-2 expression, as well as the presence of nucleobindin-2 within the plasma of rodents and humans, strongly suggests that this compound may act as a circulating regulatory factor. NUCB2 can be cleaved into three polypeptides: nesfatin-1, nesfatin-2, and nesfatin-3. Anorexigenic effects have been described for nesfatin-1 that plays an important role in hypothalamic pathways regulating food intake and energy homeostasis and acting in a leptin-independent manner. Nesfatin-1 may also exert hypertensive roles and modulate blood pressure through directly acting on peripheral arterial resistance. Nucleobindin-2 is therefore reported to reduce food intake after administration, mostly refering to the cleaved peptide nesfatin-1. Excess nucleobindin-2/nesfatin-1 in the brain leads to a loss of appetite, lack of the protein in the brain leads to an increase of appetite.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:<1EU/µg
- Gene ID:P81117
- Protein Synonyms:Prepronesfatin|Nesfatin|DNA-binding Protein NEFA|Nucb2
- Protein/Peptide Name:NUCB2
- Purity:90%
- Molecular Weight:~55kDa (SDS-PAGE)
- Sequence:25-420
- Endotoxin Level:Low
- Formulation:Liquid. 0.2µm-filtered solution in PBS, pH 7.2.
- Shipping Temperature:-20 °C, Blue Ice