ANGPTL7 (Angiopoietin-like protein 7) is expressed in various tissues and regulates angiogenesis. Human ANGPTL7 was characterized as a potent target gene of the WNT/beta-catenin signaling pathway, and is a pharmacogenomics target in the fields of oncology and regenerative medicine. Overexpression of ANGPTL7 increases collagen expression and might exert a pathogenic role in glaucoma.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:<5EU/mg
- Biological Activity:Activates ERK phosphorylation in THP-1 cells.
- Gene ID:O43827
- Protein Synonyms:Angiopoietin-like Protein 7|Cornea-derived Transcript 6 Protein
- Protein/Peptide Name:ANGPTL7
- Purity:90%
- Molecular Weight:~45kDa (SDS-PAGE)
- Sequence:1-346
- Endotoxin Level:Low
- Concentration:0.5 mg/ml
- Formulation:Liquid. 0.2µm-filtered solution in PBS.
- Shipping Temperature:-20 °C, Blue Ice