Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant FABP4 (from E. coli)
Human Recombinant FABP4 (from E. coli)
Catalog # 102514-806
Supplier:  Adipogen
Small Business Enterprise
CAS Number:  
Human Recombinant FABP4 (from E. coli)
Catalog # 102514-806
Supplier:  Adipogen
Supplier Number:  AG-40A-0035-C050
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1EU/µg
  • Gene ID:
    P15090
  • Protein Synonyms:
    ALBP|Adipocyte Fatty Acid Binding Protein|Fatty Acid Binding Protein 4|Adipocyte Lipid Binding Protein
  • Protein/Peptide Name:
    FABP4
  • Purity:
    90%
  • Molecular Weight:
    ~16kDa (SDS-PAGE)
  • Sequence:
    1-132
  • Endotoxin Level:
    Low
  • Concentration:
    1mg/ml
  • Formulation:
    Liquid. 0.2µm-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 200mM NaCl.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102514-806
  • Supplier no.:
    AG40A0035C050

Specifications

About this item

FABP4 is a fatty acid binding protein in adipocytes. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABPs regulate the fatty acid uptake, transport, and metabolism. aP2, the mouse form of FABP4, seems to be central to the pathway that links obesity to insulin resistance. aP2 was shown to be an adipokine linking adipocytes to hepatic glucose production and that neutralizing secreted aP2 may represent an effective therapeutic strategy against diabetes.