Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant Resistin (from HEK293 cells)
Mouse Recombinant Resistin (from HEK293 cells)
Catalog # 102514-748
Supplier:  Adipogen
Small Business Enterprise
CAS Number:  
Mouse Recombinant Resistin (from HEK293 cells)
Catalog # 102514-748
Supplier:  Adipogen
Supplier Number:  AG-40A-0011-C050
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Size:
    50 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.1EU/µg
  • Gene ID:
    Q99P87
  • Protein Synonyms:
    Adipose Tissue-Specific Secretory Factor|FIZZ3|Found in Inflammatory Zone 3|ADSF
  • Protein/Peptide Name:
    Resistin
  • Purity:
    90%
  • Molecular Weight:
    ~15kDa (SDS-PAGE)
  • Sequence:
    21-114
  • Endotoxin Level:
    Low
  • Concentration:
    0.4mg/ml
  • Formulation:
    Liquid. 0.2µm-filtered solution in PBS, pH 7.2.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102514-748
  • Supplier no.:
    AG40A0011C050

Specifications

About this item

The adipocytokine resistin which belongs to a family of cysteine-rich C-terminal proteins known as resistin-like molecules (RELM; RELM-alpha/FIZZ 1 and RELM-beta/FIZZ 2) of FIZZ (found in inflammatory zone) are thought to be involved in inflammatory processes. Previous studies in mice showed that resistin impairs glucose tolerance and insulin action. In addition, resistin also inhibits adipogenesis in murine 3T3-L1 cells. Therefore resistin has also been proposed as an adipocyte-secreted factor thought to link obesity and T2DM.