Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Control
- Antigen Symbol:Mouse
- Clonality:Monoclonal
- Clone:15H6
- Conjugation:ATTO 647N
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Size:50 µg
- Form:Liquid
- Storage Buffer:In PBS containing 0.02% sodium azide.
- Storage Temperature:+4 °C
- Concentration:1mg/ml
- Shipping Temperature:-20° C
- Immunogen:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Tested Applications:FACS
- Cat. No.:102511-428
- Supplier no.:35B0003TSC050
Negative control in assays where mouse IgG1 (ATTO 647N)-labeled antibodies are used.
Stable for at least 1 year after receipt when stored at +4°C.
Purity: >95% (SDS-PAGE)
Type:
Antigen: Mouse
Clonality: Monoclonal
Clone: 15H6
Conjugation: ATTO 647N
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: