Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:Giantin
- Clone:TA10
- Conjugation:ATTO 488
- Host:HEK293
- ImmunoChemistry:Yes
- Isotype:IgG2 lambda
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q14789
- Antigen Synonyms:GCP372|372 kDa Golgi Complex-associated Protein|Golgin Subfamily B Member 1|Macrogolgin
- Storage Buffer:In PBS containing 0.02% sodium azide.
- Storage Temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1mg/ml
- Shipping Temperature:-20° C
- Immunogen:Human recombinant giantin.
- Cat. No.:102511-418
- Supplier no.:27B0003TDC100
Specifications
About this item
Giantin, the largest Golgi membrane protein, is C-terminally anchored to cis- and medial-Golgi membranes and forms a long ~250nm coiled-coil rod that directly interacts with microtubules in a cell-cycle regulated way. It is an excellent marker for mammalian golgi complex. It is also an antigen in chronic rheumatoid arthritis and in the autoimmune disease Sjoegren syndrome.
- Purified from HEK 293 cell culture supernatant.
Keep conjugated formats at +4°C. Stable for at least 1 year after receipt when stored at +4°C.
Type:
Antigen: Giantin
Clonality:
Clone: TA10
Conjugation: ATTO 488
Epitope:
Host: Human
Isotype: IgG2 lambda
Reactivity: Human, Mouse