Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:Obestatin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q9UBU3
- Storage Buffer:0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage Temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1mg/ml
- Shipping Temperature:-20° C
- Immunogen:Synthetic peptide corresponding to aa 76-98 (F76NAPFDVGIKLSGVQYQQHSQAL98) of human obestatin.
- Cat. No.:102514-490
- Supplier no.:AG25A0043C100
Obestatin derives like ghrelin from proghrelin, but exhibits opposite physiological effects. Obestatin decreases food intake and body weight in rodents. Moreover, obestatin also counteracts ghrelin's effect by decelerating gastric emptying and decreasing intestinal contractility. Obestatin binds to orphan G protein-coupled receptor GPR39. Amidation of the C-terminus seems to be important for obestatin's activity.
Type:
Antigen: Obestatin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse