Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Resistin-like alpha
- Antigen Symbol:RELMA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Rat
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q99P85
- Antigen Synonyms:PMNG1|HIMF|Resistin-like molecule-alpha|FIZZ1
- Storage Buffer:0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage Temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1mg/ml
- Shipping Temperature:-20° C
- Immunogen:Recombinant rat RELM-alpha.
- Cat. No.:102514-406
- Supplier no.:AG25A0011C100
Specifications
About this item
RELM-alpha is a member to the family of adipocyte-secreted proteins called adipocytokines. It is abundantly expressed in white adipose tissue but not in preadipocytes, nor 3T3-L1 adipocytes. RELM-alpha is involved in the regulation of colonic inflammation and might be a link between colonic injury, glucose tolerance, and energy intake.
Type:
Antigen: RELM-alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat