Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Caspase-1 Mouse Monoclonal Antibody [clone: Casper-1]
Anti-Caspase-1 Mouse Monoclonal Antibody [clone: Casper-1]
Catalog # 102514-354
Supplier:  Adipogen
Anti-Caspase-1 Mouse Monoclonal Antibody [clone: Casper-1]
Catalog # 102514-354
Supplier:  Adipogen
Supplier Number:  AG-20B-0042-C100

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Caspase 1
  • Antigen Symbol:
    CASP1
  • Clonality:
    Monoclonal
  • Clone:
    Casper-1
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Form:
    Liquid
  • Gene ID:
    P29452
  • Antigen Synonyms:
    Interleukin-1beta-converting Enzyme|ICE|IL-1BC|Interleukin-1beta Convertase
  • Storage Buffer:
    In PBS containing 10% glycerol and 0.02% sodium azide.
  • Storage Temperature:
    Short term: +4 °C, Long Term: –20 °C
  • Concentration:
    1mg/ml
  • Shipping Temperature:
    -20° C
  • Immunogen:
    Recombinant mouse caspase-1.
  • Cat. No.:
    102514-354
  • Supplier no.:
    AG20B0042C100

Specifications

About this item

Caspase-1 is the best-described inflammatory caspase. It processes the cytokines interleukin-1beta (IL-1beta) and IL-18 and induces pyroptotic cell death. Caspase-1 is activated by multiprotein complexes called Inflammasomes in response to numerous stimuli that are detected through distinct inflammasomes. NLRC4 responds to cytosolic flagellin, murine NLRP1b responds to anthrax lethal toxin, AIM2 responds to cytosolic DNA and NLRP3 responds to a variety of agonists including crystals.

  • Stable for at least 1 year after receipt when stored at -20°C.

Purified from concentrated hybridoma tissue culture supernatant.

Purity: >95% (SDS-PAGE)

Type:
Antigen: Caspase-1
Clonality: Monoclonal
Clone: Casper-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse