Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-BAFF-R Rat Monoclonal Antibody (ATTO 647N) [clone: 9B9]
Anti-BAFF-R Rat Monoclonal Antibody (ATTO 647N) [clone: 9B9]
Catalog # 102511-404
Supplier:  Adipogen
Anti-BAFF-R Rat Monoclonal Antibody (ATTO 647N) [clone: 9B9]
Catalog # 102511-404
Supplier:  Adipogen
Supplier Number:  AG-20B-0034TS-T100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    BAFF-R
  • Clonality:
    Monoclonal
  • Clone:
    9B9
  • Conjugation:
    ATTO 647N
  • Flow Cytometry:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Size:
    100 tests
  • Form:
    Liquid
  • Gene ID:
    Q9D8D0
  • Antigen Synonyms:
    BR3|CD268|BlySR3|TNFRSF13C|BAFF Receptor
  • Storage Buffer:
    In PBS containing 0.02% sodium azide.
  • Storage Temperature:
    Short term: +4 °C, Long Term: –20 °C
  • Concentration:
    1mg/ml
  • Shipping Temperature:
    -20° C
  • Immunogen:
    Y3 cells transfected with mouse BAFF-R.
  • Tested Applications:
    FACS, Functional Blocking
  • Cat. No.:
    102511-404
  • Supplier no.:
    20B0034TST100

Specifications

About this item

Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.

  • Purified from concentrated hybridoma tissue culture supernatant.
  • Purity: >95% (SDS-PAGE)

Keep conjugated formats at +4°C.

Type:
Antigen: BAFF-R
Clonality: Monoclonal
Clone: 9B9
Conjugation: ATTO 647N
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse