Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:TNF Receptor Superfamily Member 10c
- Antigen Symbol:TRAIL-R3
- Clonality:Monoclonal
- Clone:HS301
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:O14798
- Antigen Synonyms:TRAIL Receptor 3|TRID|DcR1|CD263|TNFRSF10C
- Storage Buffer:In PBS containing 10% glycerol and 0.02% sodium azide.
- Storage Temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1mg/ml
- Shipping Temperature:-20° C
- Immunogen:Recombinant human TRAIL-R3 (DcR1).
- Tested Applications:FACS
- Cat. No.:102514-312
- Supplier no.:AG20B0024C100
Specifications
About this item
TRAIL-R3 is a receptor for the cytotoxic ligand TRAIL. It lacks a cytoplasmic death domain and hence is not capable of inducing apoptosis. Protects cells against TRAIL mediated apoptosis by competing with TRAIL-R1 and R2 for binding to the ligand.
- Stable for at least 1 year after receipt when stored at -20°C.
Purified from concentrated hybridoma tissue culture supernatant.
Purity: >95% (SDS-PAGE)
Type:
Antigen: TRAIL-R3
Clonality: Monoclonal
Clone: HS301
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human