Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-RIG-I Mouse Monoclonal Antibody (Biotin) [clone: Alme-1]
Anti-RIG-I Mouse Monoclonal Antibody (Biotin) [clone: Alme-1]
  102514-270
 :  Adipogen
Anti-RIG-I Mouse Monoclonal Antibody (Biotin) [clone: Alme-1]
  102514-270
 :  Adipogen
 :  AG-20B-0009B-C100

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Retinoic Acid-inducible Gene 1
  • Antigen Symbol:
    RIG-1
  • Clonality:
    Monoclonal
  • Clone:
    Alme-1
  • Conjugation:
    Biotin
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Form:
    Liquid
  • Gene ID:
    O95786
  • Antigen Synonyms:
    Probable ATP-dependent RNA Helicase DDX58|DEAD-box Protein 58|RIG-1|Retinoic Acid-inducible Gene 1 Protein
  • Storage Buffer:
    In PBS containing 0.02% sodium azide.
  • Storage Temperature:
    Short term: +4 °C, Long Term: –20 °C
  • Concentration:
    0.5mg/ml
  • Shipping Temperature:
    -20° C
  • Immunogen:
    Recombinant human RIG-I (aa 201-713).
  • Cat. No.:
    102514-270
  • Supplier no.:
    AG20B0009BC100

 

 

RIG-I and MDA5 are highly conserved helicases involved in the innate immune response to virus. RIG-I is a member of the DEAD-box RNA helicases and is activated by cytoplasmic dsRNA and 5'-ppp RNA produced during the viral replication. The protein is characterized by a N-terminal region with two caspase recruitment domains (CARD) and a C-terminal region harboring potential ATP-dependent RNA helicase activity. RIG-I recruits the CARD adaptor inducing IFN-beta (Cardif) in a CARD-CARD-dependent manner resulting in NF-kappaB and IRF3 activation.

  • Stable for at least 1 year after receipt when stored at -20°C.

Purified from concentrated hybridoma tissue culture supernatant.

Purity: >95% (SDS-PAGE)

Type:
Antigen: RIG-I
Clonality: Monoclonal
Clone: Alme-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse