DLK1 (Protein delta homolog 1; Preadipocyte factor 1; Pref-1) is a regulator of adipocyte differentiation found in serum and urine. DLK1 might be a decoy ligand. Serum or plasma levels of soluble DLK1 affect negatively or positively adipogenesis and control mesenchymal cell fate. DLK1 is frequently upregulated in myelodysplastic syndrome (MDS) patients compared to non-leukemic individuals.
Type:
Antigen: DLK1
Clonality: Monoclonal
Clone: PF13-3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- 102514-162
- 102514-160
- Antigen Symbol:
- DLK1
- DLK1
- Antigen Name:
- Delta-like 1 Homolog (Drosophila)
- Delta-like 1 Homolog (Drosophila)
- Antigen Synonyms:
- Fetal Antigen 1|Pref-1|FA1|Protein Delta Homolog 1|Preadipocyte Factor 1
- Fetal Antigen 1|Pref-1|FA1|Protein Delta Homolog 1|Preadipocyte Factor 1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- PF13-3
- PF13-3
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- P80370
- P80370
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Recombinant human DLK1 (extracellular domain).
- Recombinant human DLK1 (extracellular domain).
- Form:
- Liquid
- Liquid
- Concentration:
- 1mg/ml
- 1mg/ml
- Storage Buffer:
- 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
- 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage Temperature:
- Short term: +4 °C, Long Term: –20 °C
- Short term: +4 °C, Long Term: –20 °C
- Shipping Temperature:
- -20° C
- -20° C
- Tested Applications:
- FACS
- FACS