Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:Ribosomal Protein S3
- Clonality:Monoclonal
- Clone:RP159-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P23396
- Antigen Synonyms:40S Ribosomal Protein S3|RPS3|IMR-90 Ribosomal Protein S3
- Storage Buffer:0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage Temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1mg/ml
- Shipping Temperature:-20° C
- Immunogen:Recombinant human ribosomal protein S3.
- Cat. No.:102514-102
- Supplier no.:AG20A0048C100
Specifications
About this item
RpS3 is a component of the 40S ribosomal subunit and is an an essential but previously unkown subunit of NF-kappaB involved in the regulation of key genes in rapid cellular activation responses. RpS3 interacts with nm23-H1. The expression of rpS3 reduces the secretion of MMP-9 and the invasive metastatic potential in HT1080 cells. The phosphorylated ERK is reduced by the expression of rpS3.
Type:
Antigen: Ribosomal Protein S3
Clonality: Monoclonal
Clone: RP159-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human