About this item
ANGPTL4 mainly expressed in endothelial cells (hypoxia-induced). Regulates angiogenesis and modulates tumorgenesis and directly regulates lipid, glucose, and energy metabolism. Inhibits proliferation, migration, and tubule formation of endothelial cells and reduces vascular leakage.
Type:
Antigen: ANGPTL4
Clonality: Monoclonal
Clone: Kairos-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 102514-062
- 102514-064
- Antigen Symbol:
- ANGPTL4
- ANGPTL4
- Antigen Synonyms:
- Hepatic Fibrinogen/Angiopoietin-related Protein|Fasting-induced Adipose Factor|HFARP|FIAF|Angiopoietin-like Protein 4
- Hepatic Fibrinogen/Angiopoietin-related Protein|Fasting-induced Adipose Factor|HFARP|FIAF|Angiopoietin-like Protein 4
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Kairos-1
- Kairos-1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- Q9BY76
- Q9BY76
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human ANGPTL4.
- Recombinant human ANGPTL4.
- Form:
- Liquid
- Liquid
- Concentration:
- 1mg/ml
- 1mg/ml
- Storage Buffer:
- 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
- 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage Temperature:
- Short term: +4 °C, Long Term: –20 °C
- Short term: +4 °C, Long Term: –20 °C
- Shipping Temperature:
- -20° C
- -20° C