Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD137L Mouse Monoclonal Antibody [clone: 41B436]
Anti-CD137L Mouse Monoclonal Antibody [clone: 41B436]
Supplier:  Adipogen
Certificates
Anti-CD137L Mouse Monoclonal Antibody [clone: 41B436]
Supplier:  Adipogen

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Catalog No:
  • 102514-030
  • 102514-028
  • Antigen Symbol:
  • CD137L
  • CD137L
  • Antigen Name:
  • Tumor Necrosis Factor (Ligand) Superfamily Member 9
  • Tumor Necrosis Factor (Ligand) Superfamily Member 9
  • Antigen Synonyms:
  • 4-1BBL|TNFSF9
  • 4-1BBL|TNFSF9
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 41B436
  • 41B436
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • P41273
  • P41273
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant human CD137L extracellular domain.
  • Recombinant human CD137L extracellular domain.
  • Form:
  • Liquid
  • Liquid
  • Concentration:
  • 1mg/ml
  • 1mg/ml
  • Storage Buffer:
  • 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
  • 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
  • Storage Temperature:
  • Short term: +4 °C, Long Term: –20 °C
  • Short term: +4 °C, Long Term: –20 °C
  • Shipping Temperature:
  • -20° C
  • -20° C
  • Tested Applications:
  • FACS
  • FACS

Specifications