Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-GITR Rat Monoclonal Antibody [clone: AIT 158D]
Anti-GITR Rat Monoclonal Antibody [clone: AIT 158D]
Supplier:  Adipogen
Certificates
Anti-GITR Rat Monoclonal Antibody [clone: AIT 158D]
Supplier:  Adipogen
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 102513-974
  • 102513-972
  • Antigen Symbol:
  • GITR
  • GITR
  • Antigen Name:
  • Tumor necrosis Factor Receptor superfamily member 18
  • Tumor necrosis Factor Receptor superfamily member 18
  • Antigen Synonyms:
  • CD357|TNFRSF18|Activation-inducible TNFR Family Receptor|Glucocorticoid-induced TNFR-related Protein|AITR|Tumor Necrosis Factor Receptor Superfamily Member 18
  • CD357|TNFRSF18|Activation-inducible TNFR Family Receptor|Glucocorticoid-induced TNFR-related Protein|AITR|Tumor Necrosis Factor Receptor Superfamily Member 18
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • AIT 158D
  • AIT 158D
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • Q9Y5U5
  • Q9Y5U5
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ELISA:
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant human GITR.
  • Recombinant human GITR.
  • Form:
  • Liquid
  • Liquid
  • Concentration:
  • 1mg/ml
  • 1mg/ml
  • Storage Buffer:
  • 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
  • 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
  • Storage Temperature:
  • Short term: +4 °C, Long Term: –20 °C
  • Short term: +4 °C, Long Term: –20 °C
  • Shipping Temperature:
  • -20° C
  • -20° C
  • Tested Applications:
  • FACS
  • FACS

Specifications