Fraser Broth, Modified, Hardy Diagnostics
89407-598
Product available on GSA Advantage®
:
- Pack type:Tube
- Size:10 mL
- Storage Temperature:2-8°C
- Cat. No.:89407-598
- Supplier no.:K98
For the selective enrichment of Listeria species (without ferric ammonium citrate).