Order Entry
Puerto Rico
ContactUsLinkComponent
Tryptic Soy Agar (TSA) with Lecithin and Tween Deep, 20×125 mm Pour Tube, Hardy Diagnostics
Tryptic Soy Agar (TSA) with Lecithin and Tween Deep, 20×125 mm Pour Tube, Hardy Diagnostics
Catalog # 89408-036
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Tryptic Soy Agar (TSA) with Lecithin and Tween Deep, 20×125 mm Pour Tube, Hardy Diagnostics
Catalog # 89408-036
Supplier:  Hardy Diagnostics
Supplier Number:  Q13
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Pour tube
  • Size:
    20 x 125 mm
  • Storage Temperature:
    2-8°C
  • Minimum Order Quantity:
    5
  • Cat. No.:
    89408-036
  • Supplier no.:
    Q13

Specifications

About this item

For the cultivation of microorganisms

  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications

CRITERION™ Dehydrated Culture Media and container.

Certifications: BSE Free