Rappaport-Vassiliadis (MSRV) Medium Base, Semisolid Modification, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
Certificates
About this item
CRITERION™ Rappaport-Vassiliadis (MSRV) Medium Base, along with novobiocin, is used in the detection and isolation of motile Salmonella spp. in feces and food products
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Sodium Novobiocin, product #SR181E, is required for this medium. Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
- Storage Temperature:–2-30°C