Order Entry
Puerto Rico
ContactUsLinkComponent
CRITERION™ Peptone Water Lecithin and Tween®80
CRITERION™ Peptone Water Lecithin and Tween®80
Supplier:  Hardy Diagnostics
Certificates
undefined
CRITERION™ Peptone Water Lecithin and Tween®80
Supplier:  Hardy Diagnostics
Certificates

Specifications

  • Storage Temperature:
    2...8 °C
  • pH:
    7.2 ±0.2 at 25 °C

Specifications