Order Entry
Puerto Rico
ContactUsLinkComponent
Mycoplasma Agar Base, CRITERION™, Hardy Diagnostics
Mycoplasma Agar Base, CRITERION™, Hardy Diagnostics
Catalog # 89406-742
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Mycoplasma Agar Base, CRITERION™, Hardy Diagnostics
Catalog # 89406-742
Supplier:  Hardy Diagnostics
Supplier Number:  C8281
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Bottle, wide mouth
  • Size:
    500 g
  • Storage Temperature:
    –2-30°C
  • Cat. No.:
    89406-742
  • Supplier no.:
    C8281

Specifications

About this item

CRITERION™ Mycoplasma Agar Base is recommended for the cultivation of Mycoplasma spp

  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications

Dehydratedculture media with engineered container.

Certifications: BSE Free