Mueller Hinton Broth, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
Certificates
About this item
CRITERION™ Mueller Hinton Broth is recommended for use in the cultivation of a wide variety of microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Mueller Hinton Broth is also recommended for antimicrobial susceptibility testing of aerobic microorganisms by broth dilution methods. Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
- Storage Temperature:–2-30°C