L-Arabinose, CRITERION™, Hardy Diagnostics
Catalog # 89406-550
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Zip-pouch, Mylar®
- Size:100 g
- Storage Temperature:–2-30°C
- Cat. No.:89406-550
- Supplier no.:C7840
Specifications
About this item
CRITERION™ L-Arabinose is primarily used in preparing microbiological culture media
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
For fermentation tests it is usually added at a final concentration of 0.5 to 1.0%. Dehydratedculture media with engineered container.
Certifications: BSE Free