Brilliant Green Bile Broth with MUG, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
CRITERION™ Brilliant Green Bile Broth with MUG is a selective medium recommended for the detection of coliforms in water, sewage, dairy products and other samples
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
The addition of 4-methylumbilliferyl-beta-D-glucuronic acid (MUG) reagent detects Escherichia coli. Dehydratedculture media with engineered container.
: BSE Free
- Storage Temperature:–2-30°C