Order Entry
Puerto Rico
ContactUsLinkComponent
 
Vogel and Johnson Agar, Hardy Diagnostics
  89407-208
Product available on GSA Advantage®
 :  
undefined
Vogel and Johnson Agar, Hardy Diagnostics
  89407-208
 :  G193
Product available on GSA Advantage®
 :  

 

  • Pack type:
    Plate
  • Size:
    15 x 100 mm
  • Storage Temperature:
    2-8°C
  • Cat. No.:
    89407-208
  • Volume:
    19 ml
  • Supplier no.:
    G193

 

 

For the selective isolation of coagulase-positive, mannitol-positive Staphylococcus aureus bacteria.