Universal Beer Agar, Hardy Diagnostics
Catalog # 89407-202
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Plate
- Description:Universal Beer Agar with Cyclohexamide (actidione)
- Size:15 x 60 mm
- Storage Temperature:2-8°C
- Minimum Order Quantity:10
- Cat. No.:89407-202
- Volume:15 ml
- Supplier no.:G182
Specifications
About this item
For the selective isolation and cultivation of microorganisms significant in the brewing industry.