Order Entry
Puerto Rico
ContactUsLinkComponent
Tryptic Soy Broth (TSB), Hardy Diagnostics
Tryptic Soy Broth (TSB), Hardy Diagnostics
  89408-056
 :  
undefined
Tryptic Soy Broth (TSB), Hardy Diagnostics
  89408-056
 :  U8210
 :  

 

  • Pack type:
    Serum vial, septum-sealed, 20 ml
  • Description:
    USP
  • Size:
    50/pack
  • Storage Temperature:
    –2-30°C
  • Cat. No.:
    89408-056
  • Volume:
    10 ml
  • Supplier no.:
    U8210