Thioglycollate Broth, Hardy Diagnostics
Catalog # 89407-972
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:without Indicator with Hemin and Vitamin K
- Size:100 mL
- Storage Temperature:–2-30°C
- Cat. No.:89407-972
- Supplier no.:U43
Specifications
About this item
For the cultivation of microorganisms and USP sterility procedures.