Starch Agar, Hardy Diagnostics
Catalog # 89407-252
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Plate
- Size:15 x 100 mm
- Storage Temperature:2-8°C
- Minimum Order Quantity:10
- Cat. No.:89407-252
- Volume:18 ml
- Supplier no.:G294
Specifications
About this item
Starch Agar is recommended for the use in the determination of starch hydrolysis by microorganisms.