Rappaport-Vassiliadis Broth, USP, Hardy Diagnostics
89407-854
Product available on GSA Advantage®
:
- Pack type:Tube
- Size:10 mL
- Storage Temperature:2-8°C
- Cat. No.:89407-854
- Supplier no.:K246
Recommended for the selective enrichment of Salmonella species from food samples, and conforms to the Harmonized USP/EP/JP requirements.