Order Entry
Puerto Rico
ContactUsLinkComponent
 
Oxford Medium, Modified, Hardy Diagnostics
Catalog # 89407-326
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Oxford Medium, Modified, Hardy Diagnostics
Catalog # 89407-326
Supplier:  Hardy Diagnostics
Supplier Number:  G46
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Plate
  • Size:
    15 x 100 mm
  • Storage Temperature:
    2-8°C
  • Cat. No.:
    89407-326
  • Volume:
    18 ml
  • Supplier no.:
    G46

Specifications

About this item

For the selective isolation and differentiation of Listeria monocytogenes .