ContactUsLinkComponent
SpotDrops™ Nitrate Reagent A, Sulfanilic Acid, Hardy Diagnostics
  89407-776
Product available on GSA Advantage®
 :  
undefined
SpotDrops™ Nitrate Reagent A, Sulfanilic Acid, Hardy Diagnostics
  89407-776
 :  Z71
Product available on GSA Advantage®
 :  
  • Pack type:
    LDPE dropping bottle
  • Size:
    15 ml
  • Storage Temperature:
    –2-30°C
  • Cat. No.:
    89407-776
  • Supplier no.:
    Z71
For use in determining the nitrate reduction reaction of bacteria, other than Mycobacterium species.