Ninhydrin Reagent, Hardy Diagnostics
Catalog # 89407-768
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Glass dropping bottle, amber
- Size:15 ml
- Storage Temperature:–2-30°C
- Cat. No.:89407-768
- Supplier no.:Z70
Specifications
About this item
For use in rapid determination of sodium hippurate hydrolysis reactions in bacteria.
Useful for the presumptive identification of Campylobacter, Listeria, and Gardnerella vaginella.