MYP (Mannitol Yolk Polymy x in) Agar, Hardy Diagnostics
Catalog # 89407-168
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Plate
- Size:15 x 100 mm
- Storage Temperature:2-8°C
- Cat. No.:89407-168
- Volume:18 ml
- Supplier no.:G147
Specifications
About this item
For use in the enumeration of Bacillus cereus from foods.