Order Entry
Puerto Rico
ContactUsLinkComponent
M-El Agar, Hardy Diagnostics
M-El Agar, Hardy Diagnostics
Catalog # 89407-150
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
M-El Agar, Hardy Diagnostics
Catalog # 89407-150
Supplier:  Hardy Diagnostics
Supplier Number:  G124
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Plate
  • Size:
    15 x 60 mm
  • Storage Temperature:
    2-8°C
  • Minimum Order Quantity:
    10
  • Cat. No.:
    89407-150
  • Volume:
    11 ml
  • Supplier no.:
    G124

Specifications

About this item

For the selective chromogenic detection and enumeration of Enterococci in water by the single-step membrane filtration technique.