m FC Media, Hardy Diagnostics
For detection of fecal coliforms in water samples by membrane filtration and elevated temperature
Contains 1% Rosolic Acid.
- Storage Temperature:2-8°C
For detection of fecal coliforms in water samples by membrane filtration and elevated temperature
Contains 1% Rosolic Acid.