Order Entry
Puerto Rico
ContactUsLinkComponent
 
E. coliPRO™ O157 Latex Kit, Hardy Diagnostics
Catalog # 89407-502
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
E. coliPRO™ O157 Latex Kit, Hardy Diagnostics
Catalog # 89407-502
Supplier:  Hardy Diagnostics
Supplier Number:  PL070HD
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    50 Tests
  • Storage Temperature:
    2-8°C
  • Cat. No.:
    89407-502
  • Supplier no.:
    PL070HD

Specifications

About this item

A rapid latex agglutination test kit for the identification of the E.coli O157 serotype.

So sensitive that it requires only one single colony. Uses proprietary antibodies that eliminate the possibility of cross-reactions with Escherichia hermanii and other sorbitol-negative Escherichia species. Contains both positive and negative controls. Includes reaction cards and mixing sticks. Rapid results in just 2 minutes. 2 year shelf life.