Order Entry
Puerto Rico
ContactUsLinkComponent
A8 Agar for Mycoplasma, Hardy Diagnostics
A8 Agar for Mycoplasma, Hardy Diagnostics
Catalog # 89407-126
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
A8 Agar for Mycoplasma, Hardy Diagnostics
Catalog # 89407-126
Supplier:  Hardy Diagnostics
Supplier Number:  G02
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Plate
  • Size:
    15 x 60 mm
  • Storage Temperature:
    2-8°C
  • Cat. No.:
    89407-126
  • Supplier no.:
    G02

Specifications

About this item

For the selective isolation of Mycoplasma hominis and Ureaplasma urealyticum.