ContactUsLinkComponent
Cytophaga Broth, CRITERION™, Hardy Diagnostics
  89406-530
Product available on GSA Advantage®
 :  
undefined
Cytophaga Broth, CRITERION™, Hardy Diagnostics
  89406-530
 :  C7791
Product available on GSA Advantage®
 :  
  • Pack type:
    Bottle, wide mouth
  • Size:
    500 g
  • Storage Temperature:
    –2-30°C
  • Cat. No.:
    89406-530
  • Supplier no.:
    C7791
CRITERION™ Cytophaga Broth for the cultivation and maintenance of Cytophaga spp
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
and Flavobacterium spp. CRITERION™ Dehydratedculture media with engineered container.
 : BSE Free