ContactUsLinkComponent
XLD Agar, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
Catalog # 89406-280
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
XLD Agar, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
Catalog # 89406-280
Supplier:  Hardy Diagnostics
Supplier Number:  C7323
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Pack type:
    Bucket
  • Size:
    10 kg
  • Cat. No.:
    89406-280
  • Supplier no.:
    C7323
Specifications
About this item
CRITERION™ XLD Agar is recommended for use as a selective and differential medium for the isolation of gram-negative enteric pathogens such as Salmonella and Shigella from fecal and other specimens.
CRITERION™ Dehydrated Culture Media and container.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
CRITERION™ XLD Agar is recommended for use as a selective and differential medium for the isolation of gram-negative enteric pathogens such as Salmonella and Shigella from fecal and other specimens.
Certifications: BSE Free