Staphylococcus 110 Agar, CRITERION™, Hardy Diagnostics
CRITERION™ Staphylococcus Medium 110 is used for isolating and differentiating pathogenic staphylococci based on fermentation, pigment formation and gelatinase activity
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
- Storage Temperature:–2-30°C