SIM Medium, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
About this item
CRITERION™ SIM Medium is recommended for the differentiation of gram-negative enteric bacilli, such as Salmonella and Shigella, on the basis of sulfide production, indole formation, and motility
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ SIM Medium is recommended for the differentiation of gram-negative enteric bacilli, such as Salmonella and Shigella, on the basis of sulfide production, indole formation, and motility
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 89406-038
- 89406-040
- 89406-042
- 89406-044
- Storage Temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Specifications
Product Family Options
Product Information
- SizeFinal volumePack typeAvailabilityPrice
- Supplier Number: C6940Specifications:Product available on GSA Advantage®
More
Specifications
Cat. No.89406-038Storage Temperature–2-30°CProduct available on GSA Advantage® - Supplier Number: C6941Specifications:Product available on GSA Advantage®
More
Specifications
Cat. No.89406-040Storage Temperature–2-30°CProduct available on GSA Advantage® - Supplier Number: C6942Specifications:Product available on GSA Advantage®
More
Specifications
Cat. No.89406-042Storage Temperature–2-30°CProduct available on GSA Advantage® - Supplier Number: C6943Specifications:Product available on GSA Advantage®
More
Specifications
Cat. No.89406-044Storage Temperature–2-30°CProduct available on GSA Advantage®