R2A Agar, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
Certificates
About this item
CRITERION™ R2A Agar is primarily for use in the cultivation and enumeration of heterotrophic microorganisms in treated potable water
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
- Storage Temperature:–2-30°C