CRITERION™ Proteose Peptone #3, Hardy Diagnostics
Supplier: Hardy Diagnostics
Certificates
About this item
CRITERION™ Proteose Peptone #3 is frequently used as a nitrogen source in media formulations for culturing nutritionally fastidious microorganisms.
- Wide mouth opening
- Reduces dust and allows easy scooping
- Convenient handgrip
- 'No slip' finger holds
- Stackable containers
- High solubility; dissolves quickly
- Custom and bulk products available
Dehydratedculture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
Certifications: BSE Free
Specifications
- Storage Temperature:–2-30°C
Specifications
Related Information
Certificates
Search by Lot NumberHardy Diagnostics Criterion Dehydrated Culture Media