Oxbile, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
Certificates
About this item
CRITERION™ Oxbile is prepared specifically for differentiation of bile tolerant microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
- Storage Temperature:–2-30°C
Specifications
Related Information
Certificates
Search by Lot NumberHardy Diagnostics Criterion Dehydrated Culture Media