mTEC Agar, CRITERION™, Hardy Diagnostics
Supplier: Hardy Diagnostics
Certificates
About this item
CRITERION™ mTEC Agar is recommended for the differentiation and enumeration of thermotolerant Escherichia coli in water by membrane filtration and an in situ urease test
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
- Storage Temperature:–2-30°C