Motility Med S Base, CRITERION™, Hardy Diagnostics
CRITERION™ Motility Med S Base for the detection of bacterial motility
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
TTC Solution (1%), product 89404-940, is required for this medium. Dehydratedculture media with engineered container.
- Storage Temperature:–2-30°C