Order Entry
Puerto Rico
ContactUsLinkComponent
CRITERION™ Modified Buffered Peptone Water
CRITERION™ Modified Buffered Peptone Water
Supplier:  Hardy Diagnostics
Certificates
undefined
CRITERION™ Modified Buffered Peptone Water
Supplier:  Hardy Diagnostics
Certificates

Specifications

  • Storage Temperature:
    2...30 °C
  • pH:
    7.2 ±0.2 at 25 °C

Specifications